accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 100-41-4

H3C

CAS RN 100-41-4

IUPAC Name: ethylbenzene
Synonyms: ethylbenzene etilbenzene ethylbenzene (substance) etylobenzen ethylenzene ccris 916 ethylbenzeen aethylbenzol hsdb 84
Molecular Weight (g/mol): 106.17
Molecular Formula: C8H10
InChi Key: YNQLUTRBYVCPMQ-UHFFFAOYSA-N
SMILES: CCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (12)
  • (7)
Quantity 
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (3)
  • (7)
Boiling Point 
  • (3)
  • (1)
  • (1)
  • (7)
Grade 
  • (1)
  • (5)
  • (2)

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (7)
  • (3)
  • (12)
  • (7)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (7)
  • (3)
  • (1)
  • (1)
  • (7)
  • (1)
  • (5)
  • (2)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.