CAS RN 1000-86-8
CAS RN 1000-86-8
IUPAC Name:
2,4-dimethylpenta-1,3-diene
Synonyms:
1,3-pentadiene, 2,4-dimethyl-
Molecular Weight (g/mol):
96.17
Molecular Formula:
C7H12
InChi Key:
CMSUNVGIWAFNBG-UHFFFAOYSA-N
SMILES:
CC(C)=CC(C)=C
1
–
2
of
2
results
1
2,4-Dimethyl-1,3-pentadiene, 95%, Thermo Scientific Chemicals
CAS: 1000-86-8 Molecular Formula: C7H12 Molecular Weight (g/mol): 96.173 MDL Number: MFCD00008903 InChI Key: CMSUNVGIWAFNBG-UHFFFAOYSA-N PubChem CID: 66080 IUPAC Name: 2,4-dimethylpenta-1,3-diene SMILES: CC(=CC(=C)C)C
2
2,4-Dimethyl-1,3-pentadiene, 98%, Thermo Scientific Chemicals
CAS: 1000-86-8 Molecular Formula: C7H12 Molecular Weight (g/mol): 96.17 MDL Number: MFCD00008903 InChI Key: CMSUNVGIWAFNBG-UHFFFAOYSA-N Synonym: 2,4-dimethyl-1,3-pentadiene,1,1,3-trimethylbutadiene,1,3-pentadiene, 2,4-dimethyl,acmc-20aoci,1,3-trimethylbutadiene,1,3-pentadiene,2,4-dimethyl,cmsunvgiwafnbg-uhfffaoysa,ch3 2c=chc ch3 =ch2,1,3-pentadiene, 2,4-dimethyl-8ci 9ci PubChem CID: 66080 IUPAC Name: 2,4-dimethylpenta-1,3-diene SMILES: CC(=CC(=C)C)C