accessibility menu, dialog, popup

Test

CAS RN 1000339-98-9

NHNFFF

CAS RN 1000339-98-9

IUPAC Name: 4-(trifluoromethyl)-1H-indazole
Synonyms: 4-(trifluoromethyl)-1H-indazole
Molecular Weight (g/mol): 186.14
Molecular Formula: C8H5F3N2
InChi Key: KCWIWQGNCLKDJZ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C2C=NNC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results