man

Test

CAS RN 10009-20-8

HNOFFFOHONH2

CAS RN 10009-20-8

IUPAC Name: 2-amino-6-(2,2,2-trifluoroacetamido)hexanoic acid
Synonyms: 2-amino-6-(2,2,2-trifluoroacetamido)hexanoic acid
Molecular Weight (g/mol): 242.2
Molecular Formula: C8H13F3N2O3
InChi Key: PZZHRSVBHRVIMI-UHFFFAOYNA-N
SMILES: NC(CCCCNC(=O)C(F)(F)F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results