accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 100130-32-3

HHBrBrH2NNH2NSNH2NH

CAS RN 100130-32-3

IUPAC Name: dihydrogen ({2-[(diaminomethylidene)amino]ethyl}sulfanyl)methanimidamide dibromide
Synonyms: dihydrogen {2-[(diaminomethylidene)amino]ethyl}sulfanylmethanimidamide dibromide
Molecular Weight (g/mol): 323.05
Molecular Formula: C4H13Br2N5S
InChi Key: GPWJSTKHQMIXCA-UHFFFAOYSA-N
SMILES: [H+].[H+].[Br-].[Br-].NC(N)=NCCSC(N)=N
Quantity 
  • (1)
  • (1)
1x1 image pixel for data collection
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.