accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1002-69-3

H3CCl

CAS RN 1002-69-3

IUPAC Name: 1-chlorodecane
Synonyms: 1-chlorodecane
Molecular Weight (g/mol): 176.73
Molecular Formula: C10H21Cl
InChi Key: ZTEHOZMYMCEYRM-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCl
1x1 image pixel for data collection
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.