man

Test

CAS RN 10022-31-8

BaONOOONOO

CAS RN 10022-31-8

IUPAC Name: barium(2+) dinitrate
Synonyms: barium nitrate
Molecular Weight (g/mol): 261.34
Molecular Formula: BaN2O6
InChi Key: IWOUKMZUPDVPGQ-UHFFFAOYSA-N
SMILES: [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (10)
  • (1)
  • (1)
Grade 
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
Physical Form 
  • (9)
  • (2)
  • (4)
Percent Purity 
  • (3)
  • (7)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (6)
  • (7)
  • (13)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (10)
  • (1)
  • (1)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (9)
  • (2)
  • (4)
  • (3)
  • (7)
  • (1)
  • (2)
  • (2)