accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10025-65-7

ClClPt

CAS RN 10025-65-7

IUPAC Name: platinum(2+) dichloride
Synonyms: platinum(II) chloride
Molecular Weight (g/mol): 265.98
Molecular Formula: Cl2Pt
InChi Key: CLSUSRZJUQMOHH-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Pt++]
Percent Purity 
  • (2)
  • (4)
  • (2)
  • (2)
Quantity 
  • (4)
  • (1)
  • (1)
  • (4)
Physical Form 
  • (2)
  • (6)
Grade 
  • (2)

Narrow Results

Narrow Results

  • (4)
  • (6)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (4)
  • (2)
  • (6)
  • (2)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.