accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10031-20-6

H2OH2OH2OH2OBrBrMn

CAS RN 10031-20-6

IUPAC Name: dibromomanganese tetrahydrate
Synonyms: manganese(II) bromide tetrahydrate
Molecular Weight (g/mol): 286.81
Molecular Formula: Br2H8MnO4
InChi Key: HHDPJRSXPOGIOP-UHFFFAOYSA-L
SMILES: O.O.O.O.Br[Mn]Br
Percent Purity 
  • (4)
  • (2)
Physical Form 
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.