accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10036-64-3

OHNOHOHOOHHOCH3

CAS RN 10036-64-3

IUPAC Name: N-[(2S,3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
Synonyms: acetyl glucosamine
Molecular Weight (g/mol): 221.21
Molecular Formula: C8H15NO6
InChi Key: OVRNDRQMDRJTHS-PVFLNQBWSA-N
SMILES: CC(=O)N[C@H]1[C@@H](O)O[C@H](CO)[C@@H](O)[C@@H]1O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.