man

Test

CAS RN 10039-54-0

NH3HONH3HOOSOOO

CAS RN 10039-54-0

IUPAC Name: bis(hydroxyazanium) sulfate
Synonyms: hydroxylammonium sulfate
Molecular Weight (g/mol): 164.13
Molecular Formula: H8N2O6S
InChi Key: VGYYSIDKAKXZEE-UHFFFAOYSA-L
SMILES: [NH3+]O.[NH3+]O.[O-]S([O-])(=O)=O
Physical Form 
  • (4)
  • (3)
  • (1)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (4)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)