accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10043-27-3

TbONOOONOOONOO

CAS RN 10043-27-3

IUPAC Name: terbium(3+) trinitrate
Synonyms: terbium(3+) trinitrate
Molecular Weight (g/mol): 344.94
Molecular Formula: N3O9Tb
InChi Key: YJVUGDIORBKPLC-UHFFFAOYSA-N
SMILES: [Tb+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Grade 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.