man

Test

CAS RN 10049-05-5

ClClCr

CAS RN 10049-05-5

IUPAC Name: λ²-chromium(2+) dichloride
Synonyms: chromium(II) chloride
Molecular Weight (g/mol): 122.9
Molecular Formula: Cl2Cr
InChi Key: XBWRJSSJWDOUSJ-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Cr++]
Percent Purity 
  • (2)
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (2)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (8)
  • (3)