accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 100524-07-0

NH2NHONH2

CAS RN 100524-07-0

IUPAC Name: 3-amino-N'-hydroxybenzene-1-carboximidamide
Synonyms: 3-amino-N'-hydroxybenzenecarboximidamide
Molecular Weight (g/mol): 151.17
Molecular Formula: C7H9N3O
InChi Key: OPGWBTKZYLRPRW-UHFFFAOYSA-N
SMILES: NC(=NO)C1=CC(N)=CC=C1
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.