accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 100564-78-1

CH3H3CCH3OOHNOHO

CAS RN 100564-78-1

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-4-phenylbutanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-4-phenylbutanoic acid
Molecular Weight (g/mol): 279.34
Molecular Formula: C15H21NO4
InChi Key: MCODLPJUFHPVQP-LBPRGKRZSA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CCC1=CC=CC=C1)C(O)=O
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.