accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 100828-16-8

HOONPOHOHOHN

CAS RN 100828-16-8

IUPAC Name: 4-(3-phosphonopropyl)piperazine-2-carboxylic acid
Synonyms: 4-(3-phosphonopropyl)piperazine-2-carboxylic acid
Molecular Weight (g/mol): 252.21
Molecular Formula: C8H17N2O5P
InChi Key: CUVGUPIVTLGRGI-UHFFFAOYNA-N
SMILES: OC(=O)C1CN(CCCP(O)(O)=O)CCN1
Quantity 
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.