accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10083-24-6

OHOHHOHO

CAS RN 10083-24-6

IUPAC Name: 5-[(1E)-2-(3,4-dihydroxyphenyl)ethenyl]benzene-1,3-diol
Synonyms: piceatannol 3-hydroxyresveratol 3,3',4'5-tetrahydroxystilbene 3,5,3',4'-tetrahydroxystilbene
Molecular Weight (g/mol): 244.25
Molecular Formula: C14H12O4
InChi Key: CDRPUGZCRXZLFL-OWOJBTEDSA-N
SMILES: OC1=CC(\C=C\C2=CC=C(O)C(O)=C2)=CC(O)=C1
Molecular Weight (g/mol) 
  • (4)
  • (1)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (4)
Grade 
  • (4)

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (4)
1x1 image pixel for data collection
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.