accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1009816-48-1

H3CHNNOSOHOHOH

CAS RN 1009816-48-1

IUPAC Name: 2-(ethylamino)-5-(hydroxymethyl)-3aH,5H,6H,7H,7aH-pyrano[3,2-d][1,3]thiazole-6,7-diol
Synonyms: 2-(ethylamino)-5-(hydroxymethyl)-3aH,5H,6H,7H,7aH-pyrano[3,2-d][1,3]thiazole-6,7-diol
Molecular Weight (g/mol): 248.3
Molecular Formula: C9H16N2O4S
InChi Key: PPAIMZHKIXDJRN-UHFFFAOYNA-N
SMILES: CCNC1=NC2C(OC(CO)C(O)C2O)S1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Color 
  • (4)
Grade 
  • (4)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.