man

Test

CAS RN 101-14-4

NH2H2NClCl

CAS RN 101-14-4

IUPAC Name: 4-[(4-amino-3-chlorophenyl)methyl]-2-chloroaniline
Synonyms: bis amine quodorole curalin M MOCA cyanaset bisamine curene 442 cuamine MT millionate M cuamine M
Molecular Weight (g/mol): 267.15
Molecular Formula: C13H12Cl2N2
InChi Key: IBOFVQJTBBUKMU-UHFFFAOYSA-N
SMILES: NC1=CC=C(CC2=CC=C(N)C(Cl)=C2)C=C1Cl
For Use With (Equipment) 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)