accessibility menu, dialog, popup

Test

CAS RN 101-26-8

BrH3CNCH3OONH3C

CAS RN 101-26-8

IUPAC Name: 3-[(dimethylcarbamoyl)oxy]-1-methylpyridin-1-ium bromide
Synonyms: pyridostigmine bromide
Molecular Weight (g/mol): 261.12
Molecular Formula: C9H13BrN2O2
InChi Key: VNYBTNPBYXSMOO-UHFFFAOYSA-M
SMILES: [Br-].CN(C)C(=O)OC1=CC=C[N+](C)=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Color 
  • (2)
Grade 
  • (2)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.