accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 101-59-7

NH2SNOO

CAS RN 101-59-7

IUPAC Name: 4-[(4-nitrophenyl)sulfanyl]aniline
Synonyms: 4-[(4-nitrophenyl)sulfanyl]aniline
Molecular Weight (g/mol): 246.28
Molecular Formula: C12H10N2O2S
InChi Key: ZBPKGHOGUVVDLF-UHFFFAOYSA-N
SMILES: NC1=CC=C(SC2=CC=C(C=C2)[N+]([O-])=O)C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.