accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 101-61-1

H3CNCH3NCH3H3C

CAS RN 101-61-1

IUPAC Name: 4-{[4-(dimethylamino)phenyl]methyl}-N,N-dimethylaniline
Synonyms: tetra-base michler ketone, reduced reduced michler's ketone methane base michler's hydride reduced michler ketone michler's ketone, reduced baze michlerova tetrabase michler methane
Molecular Weight (g/mol): 254.38
Molecular Formula: C17H22N2
InChi Key: JNRLEMMIVRBKJE-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(CC2=CC=C(C=C2)N(C)C)C=C1
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.
1x1 image pixel for data collection