accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 101-75-7

HNNN

CAS RN 101-75-7

IUPAC Name: N-phenyl-4-(2-phenyldiazen-1-yl)aniline
Synonyms: N-phenyl-4-(2-phenyldiazen-1-yl)aniline
Molecular Weight (g/mol): 273.34
Molecular Formula: C18H15N3
InChi Key: VXLFYNFOITWQPM-UHFFFAOYSA-N
SMILES: N(C1=CC=CC=C1)C1=CC=C(C=C1)N=NC1=CC=CC=C1
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.