man

Test

CAS RN 10112-91-1

ClClHgHg

CAS RN 10112-91-1

IUPAC Name: dimercuranebis(ylium) dichloride
Synonyms: mercury(I) chloride
Molecular Weight (g/mol): 472.08
Molecular Formula: Cl2Hg2
InChi Key: ZOMNIUBKTOKEHS-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Hg+][Hg+]
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (4)
  • (1)
  • (3)
Grade 
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)