accessibility menu, dialog, popup

Test

CAS RN 101199-38-6

NSSOOONa

CAS RN 101199-38-6

IUPAC Name: sodium 3-(10H-phenothiazin-10-yl)propane-1-sulfonate
Synonyms: sodium 3-(phenothiazin-10-yl)propane-1-sulfonate
Molecular Weight (g/mol): 343.39
Molecular Formula: C15H14NNaO3S2
InChi Key: VKGYKXFNSKEHED-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)CCCN1C2=CC=CC=C2SC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results