accessibility menu, dialog, popup

Test

CAS RN 101303-98-4

CH3OONHNClH2NHCl

CAS RN 101303-98-4

IUPAC Name: 4-amino-N-{1-azabicyclo[2.2.2]octan-3-yl}-5-chloro-2-methoxybenzamide hydrochloride
Synonyms: zacopride hydrochloride
Molecular Weight (g/mol): 346.25
Molecular Formula: C15H21Cl2N3O2
InChi Key: ITXVOUSORXSTQH-UHFFFAOYNA-N
SMILES: Cl.COC1=CC(N)=C(Cl)C=C1C(=O)NC1CN2CCC1CC2
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results