man

Test

CAS RN 101366-51-2

CH3SClOO

CAS RN 101366-51-2

IUPAC Name: 4'-methyl-[1,1'-biphenyl]-4-sulfonyl chloride
Synonyms: 4'-methyl-[1,1'-biphenyl]-4-sulfonyl chloride
Molecular Weight (g/mol): 266.74
Molecular Formula: C13H11ClO2S
InChi Key: DBYXACYHGMSPMQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=CC=C(C=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (4)
  • (2)
Quantity 
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)