accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10147-37-2

H3CCH3SClOO

CAS RN 10147-37-2

IUPAC Name: propane-2-sulfonyl chloride
Synonyms: propane-2-sulfonyl chloride
Molecular Weight (g/mol): 142.6
Molecular Formula: C3H7ClO2S
InChi Key: DRINJBFRTLBHNF-UHFFFAOYSA-N
SMILES: CC(C)S(Cl)(=O)=O
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.