accessibility menu, dialog, popup

Test

CAS RN 10152-76-8

CH3SH2C

CAS RN 10152-76-8

IUPAC Name: 3-(methylsulfanyl)prop-1-ene
Synonyms: allyl methyl sulfide
Molecular Weight (g/mol): 88.17
Molecular Formula: C4H8S
InChi Key: NVLPQIPTCCLBEU-UHFFFAOYSA-N
SMILES: CSCC=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results