man

Test

CAS RN 10160-87-9

H3COOCH3CH

CAS RN 10160-87-9

IUPAC Name: 3,3-diethoxyprop-1-yne
Synonyms: 3,3-diethoxy-propyne
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: RGUXEWWHSQGVRZ-UHFFFAOYSA-N
SMILES: CCOC(OCC)C#C
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)