accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10172-89-1

H3COHNOHO

CAS RN 10172-89-1

IUPAC Name: (2R)-2-acetamido-3-phenylpropanoic acid
Synonyms: N-acetyl-D-phenylalanine
Molecular Weight (g/mol): 207.23
Molecular Formula: C11H13NO3
InChi Key: CBQJSKKFNMDLON-SNVBAGLBSA-N
SMILES: CC(=O)N[C@H](CC1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.