accessibility menu, dialog, popup

Test

CAS RN 1017778-98-1

CH3OOFFF

CAS RN 1017778-98-1

IUPAC Name: 2-methoxy-6-(trifluoromethyl)benzaldehyde
Synonyms: 2-methoxy-6-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: LLBTYKHVRXDHKF-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1C=O)C(F)(F)F

 No Refinements