accessibility menu, dialog, popup

Test

CAS RN 1017779-38-2

CH3OFBrF

CAS RN 1017779-38-2

IUPAC Name: 5-(bromomethyl)-2-ethoxy-1,3-difluorobenzene
Synonyms: 5-(bromomethyl)-2-ethoxy-1,3-difluorobenzene
Molecular Weight (g/mol): 251.07
Molecular Formula: C9H9BrF2O
InChi Key: SFMOQKHETBGLAR-UHFFFAOYSA-N
SMILES: CCOC1=C(F)C=C(CBr)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results