accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1017779-42-8

FOFFBr

CAS RN 1017779-42-8

IUPAC Name: 1-(bromomethyl)-2-(difluoromethoxy)-4-fluorobenzene
Synonyms: 1-(bromomethyl)-2-(difluoromethoxy)-4-fluorobenzene
Molecular Weight (g/mol): 255.03
Molecular Formula: C8H6BrF3O
InChi Key: IGODPIYHAUGSCK-UHFFFAOYSA-N
SMILES: FC(F)OC1=C(CBr)C=CC(F)=C1
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.