accessibility menu, dialog, popup

Test

CAS RN 10192-85-5

KOOH2C

CAS RN 10192-85-5

IUPAC Name: potassium prop-2-enoate
Synonyms: potassium polyacrylate
Molecular Weight (g/mol): 110.15
Molecular Formula: C3H3KO2
InChi Key: ZUBIJGNKOJGGCI-UHFFFAOYSA-M
SMILES: [K+].[O-]C(=O)C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results