accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10199-53-8

CH3NNOOH

CAS RN 10199-53-8

IUPAC Name: 1-methyl-5-phenyl-1H-pyrazole-3-carboxylic acid
Synonyms: 1-methyl-5-phenylpyrazole-3-carboxylic acid
Molecular Weight (g/mol): 202.21
Molecular Formula: C11H10N2O2
InChi Key: ZDUDGEACIDDEPL-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1C1=CC=CC=C1)C(O)=O
Quantity 
  • (2)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.