accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 102-24-9

H3COBOBOCH3OBOH3CO

CAS RN 102-24-9

IUPAC Name: trimethoxy-1,3,5,2,4,6-trioxatriborinane
Synonyms: boroxin, trimethoxy-
Molecular Weight (g/mol): 173.53
Molecular Formula: C3H9B3O6
InChi Key: ZFMOJHVRFMOIGF-UHFFFAOYSA-N
SMILES: COB1OB(OC)OB(OC)O1
Quantity 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.