man

Test

CAS RN 102-54-5

Fe

CAS RN 102-54-5

IUPAC Name: λ²-iron(2+) bis(cyclopenta-2,4-dien-1-ide)
Synonyms: ferrocene
Molecular Weight (g/mol): 186.04
Molecular Formula: C10H10Fe
InChi Key: KTWOOEGAPBSYNW-UHFFFAOYSA-N
SMILES: [Fe++].[CH-]1C=CC=C1.[CH-]1C=CC=C1
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)