accessibility menu, dialog, popup

Test

CAS RN 102-85-2

CAS RN 102-85-2

IUPAC Name: tributyl phosphite
Synonyms: phosphorous acid, tributyl ester
Molecular Weight (g/mol): 250.32
Molecular Formula: C12H27O3P
InChi Key: XTTGYFREQJCEML-UHFFFAOYSA-N
SMILES: CCCCOP(OCCCC)OCCCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)