man

Test

CAS RN 10213-79-3

H2OH2OH2OH2OH2ONaNaOSiOO

CAS RN 10213-79-3

IUPAC Name: disodium oxosilanebis(olate) pentahydrate
Synonyms: sodium silicate
Molecular Weight (g/mol): 212.14
Molecular Formula: H10Na2O8Si
InChi Key: ADPGKKZKGXANON-UHFFFAOYSA-N
SMILES: O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)