accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10230-69-0

CAS RN 10230-69-0

IUPAC Name: 1-(benzylsulfanyl)propan-2-one
Synonyms: 1-(benzylsulfanyl)propan-2-one
Molecular Weight (g/mol): 180.27
Molecular Formula: C10H12OS
InChi Key: OIEDQMIEPJIRFT-UHFFFAOYSA-N
SMILES: CC(=O)CSCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.