accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10236-14-3

CAS RN 10236-14-3

IUPAC Name: ethyl (2E)-4-(diethoxyphosphoryl)but-2-enoate
Synonyms: ethyl (2E)-4-(diethoxyphosphoryl)but-2-enoate
Molecular Weight (g/mol): 250.23
Molecular Formula: C10H19O5P
InChi Key: LXLODBXSCRTXFG-BQYQJAHWSA-N
SMILES: CCOC(=O)\C=C\CP(=O)(OCC)OCC
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.