accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 102684-91-3

CAS RN 102684-91-3

IUPAC Name: 2-bromo-5-(trifluoromethyl)benzaldehyde
Synonyms: 2-bromo-5-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 253.02
Molecular Formula: C8H4BrF3O
InChi Key: CSOBJYGHQOLWOD-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(C=O)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.