accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10294-31-2

CAS RN 10294-31-2

IUPAC Name: gold(1+) iodide
Synonyms: gold monoiodide
Molecular Weight (g/mol): 323.87
Molecular Formula: AuI
InChi Key: ATGIETUGWDAYPU-UHFFFAOYSA-M
SMILES: [I-].[Au+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.