accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 103058-87-3

CAS RN 103058-87-3

IUPAC Name: 5-bromo-2-methoxypyridine-3-carbaldehyde
Synonyms: 5-bromo-2-methoxypyridine-3-carbaldehyde
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: VRNOWEKCASDTFG-UHFFFAOYSA-N
SMILES: COC1=NC=C(Br)C=C1C=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.