accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 103404-87-1

CAS RN 103404-87-1

IUPAC Name: sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
Synonyms: hydroxyethylpiperazine ethane sulfonic acid
Molecular Weight (g/mol): 260.28
Molecular Formula: C8H17N2NaO4S
InChi Key: RDZTWEVXRGYCFV-UHFFFAOYSA-M
SMILES: [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.