accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 104-01-8

CAS RN 104-01-8

IUPAC Name: 2-(4-methoxyphenyl)acetic acid
Synonyms: 4-methoxyphenylacetic acid MOPA p-methoxy-a-toluate p-methoxy-α-toluate homo-p-anisic acid homoanisate p-methoxy-α-toluic acid 2-(p-anisyl)acetic acid P-methoxyphenylacetic acid homoanisic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: NRPFNQUDKRYCNX-UHFFFAOYSA-N
SMILES: COC1=CC=C(CC(O)=O)C=C1
Boiling Point 
  • (3)
  • (4)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (4)

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (4)
  • (3)
  • (4)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.