accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 104530-79-2

CAS RN 104530-79-2

IUPAC Name: oxolan-3-amine
Synonyms: oxolan-3-amine
Molecular Weight (g/mol): 87.12
Molecular Formula: C4H9NO
InChi Key: MIPHRQMEIYLZFZ-UHFFFAOYNA-N
SMILES: NC1CCOC1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.