accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1049741-55-0

CAS RN 1049741-55-0

IUPAC Name: 2-({2-[(4-methoxyphenyl)amino]pyrimidin-4-yl}amino)ethan-1-ol hydrochloride
Synonyms: 2-({2-[(4-methoxyphenyl)amino]pyrimidin-4-yl}amino)ethanol hydrochloride
Molecular Weight (g/mol): 296.76
Molecular Formula: C13H17ClN4O2
InChi Key: QQAHYSZVJLHCNA-UHFFFAOYSA-N
SMILES: Cl.COC1=CC=C(NC2=NC=CC(NCCO)=N2)C=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.