accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 115383-22-7

CAS RN 115383-22-7

IUPAC Name: hexatriacontacyclo[11.3.54.0¹⁷,²².0²,²³.0¹⁴,¹⁸.0¹⁹,⁷⁰.0²⁰,³⁷.0²¹,³⁴.0⁸,¹⁶.0¹⁰,¹⁵.0⁹,⁶³.0³,⁷.0⁴,²⁵.0⁶,⁶².0⁵,⁴⁸.0²⁴,³³.0³¹,³⁵.0²⁷,³².0²⁶,⁴⁷.0²⁸,⁴⁶.0⁴⁵,⁵⁰.0⁴⁹,⁶¹.0⁵¹,⁵⁹.0⁴⁴,⁵².0⁶⁰,⁶⁴.0¹¹,⁶⁵.0¹²,⁶⁷.0⁵⁸,⁶⁶.0⁵⁶,⁶⁸.0³⁸,⁶⁹.0³⁹,⁵⁵.0³⁶,⁴⁰.0³⁰,⁴¹.0²⁹,⁴³.0⁴²,⁵⁴.0⁵³,⁵⁷]heptaconta-1,3,5(48),6,8(16),9,11(65),12(67),13(70),14,17(22),18,20,23,25,27(32),28,30(41),31(35),33,36,38(69),39,42,44(52),45(50),46,49(61),51(59),53(57),54,56(68),58(66),60(64),62-pentatriacontaene
Synonyms: hexatriacontacyclo[11.3.54.0¹⁷,²².0²,²³.0¹⁴,¹⁸.0¹⁹,⁷⁰.0²⁰,³⁷.0²¹,³⁴.0⁸,¹⁶.0¹⁰,¹⁵.0⁹,⁶³.0³,⁷.0⁴,²⁵.0⁶,⁶².0⁵,⁴⁸.0²⁴,³³.0³¹,³⁵.0²⁷,³².0²⁶,⁴⁷.0²⁸,⁴⁶.0⁴⁵,⁵⁰.0⁴⁹,⁶¹.0⁵¹,⁵⁹.0⁴⁴,⁵².0⁶⁰,⁶⁴.0¹¹,⁶⁵.0¹²,⁶⁷.0⁵⁸,⁶⁶.0⁵⁶,⁶⁸.0³⁸,⁶⁹.0³⁹,⁵⁵.0³⁶,⁴⁰.0³⁰,⁴¹.0²⁹,⁴³.0⁴²,⁵⁴.0⁵³,⁵⁷]heptaconta-1,3,5(48),6,8(16),9,11(65),12(67),13(70),14,17(22),18,20,23,25,27(32),28,30(41),31(35),33,36,38(69),39,42,44(52),45(50),46,49(61),51(59),53(57),54,56(68),58(66),60(64),62-pentatriacontaene rugbyballene C70 fullerene fullerene C70 fullerene 70
Molecular Weight (g/mol): 840.77
Molecular Formula: C70
InChi Key: ATLMFJTZZPOKLC-UHFFFAOYSA-N
SMILES: C12=C3C4=C5C1=C1C6=C7C8=C9C%10=C%11C%12=C%13C%14=C%15C%16=C%17C%18=C%19C%20=C%21C%22=C%19C%19=C%23C(C%11=C%19C%18=C%12%16)=C9C9=C%11C(=C2C9=C18)C1=C(C2=C%21C8=C9C(=C4C4=C9C9=C%12C(=C%17C%20=C8%12)C%15=C8C%14=C(C%12=C6C5=C4C%12=C98)C7=C%10%13)C2=C31)C%22=C%23%11
Boiling Point 
  • (7)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)

Narrow Results

Narrow Results

  • (7)
  • (7)
  • (2)
  • (2)
  • (3)
  • (7)
  • (2)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.